Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Mouse Anti-Human Granzyme A, BD Pharmingen™
Mouse Monoclonal Antibody
$188.00 - $354.00
Specifications
Antigen | Granzyme A |
---|---|
Clone | CB9 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
Granzyme A Monoclonal antibody specifically detects Granzyme A in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
Granzyme A | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
CTL tryptase; CTLA3; GZMA; GrA; HFSP; Hanukkah factor | |
Human Cytotoxic T Cell Granule Proteins | |
Affinity Chromatography |
CB9 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
3001 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title