Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PE Mouse Anti-Mouse TCR Vγ6, BD Pharmingen™

Catalog No. BDB570216 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570216 0.1 mg
BDB570217 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570216 Supplier BD Biosciences Supplier No. 570216
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR Vγ6 Monoclonal antibody specifically detects TCR Vγ6 in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vγ6
Applications Flow Cytometry
Classification Monoclonal
Clone 1C10-1F7
Concentration 0.2 mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TCR Vg6; TCR Vgamma6; TCR Vγ6; TRGV6; TcR Vγ4; Tcrg-V6; Trgv6
Host Species Mouse
Immunogen Mouse TCR Vγ6 Peptide
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 21640
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.