Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PE Mouse Anti-Mouse TCR Vδ6B, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570738
Description
TCR Vδ6B Monoclonal antibody specifically detects TCR Vδ6B in Mouse samples. It is validated for Flow cytometry
Specifications
TCR Vδ6B | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
F4.22.rMAb (also known as Anti-Vδ6 λ12) | |
PE | |
TCR Variable delta 6B; TCR Vd6b; TCR Vδ6 λ12 | |
Mouse γδ T Cell Hybridoma Cells | |
0.1 mg | |
110371 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction