Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PE Rat Anti-β-galactosidase, BD Pharmingen™

Catalog No. BDB570543 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570543 25 μg
BDB570542 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570543 Supplier BD Biosciences Supplier No. 570543
Add to Cart
Add to Cart

Rat Monoclonal Antibody

β-galactosidase Monoclonal antibody specifically detects β-galactosidase in Bacteria samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-galactosidase
Applications Flow Cytometry
Classification Monoclonal
Clone 13R4.rMAb (also known as 13R4)
Concentration 0.2 mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias beta-gal; beta-galactosidase; lacZ; lactase; β-gal; β-galactosidase
Host Species Rat
Immunogen E. coli β-galactosidase
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 945006
Target Species Bacteria
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.