Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEA-15 Antibody, PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305543PE
Description
PEA-15 Monoclonal antibody specifically detects PEA-15 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
PEA-15 | |
Monoclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
HMAT115kD, PEA-15, PED15 kDa phosphoprotein enriched in astrocytes, phosphoprotein enriched in astrocytes 15, Phosphoprotein enriched in diabetes | |
Full length human PEA-15 as expressed in and purified from E. coli. | |
0.1 mL | |
Apoptosis, Cell Biology, Cellular Markers, Phospho Specific | |
8682 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction