Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEA15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584852 Supplier Invitrogen™ Supplier No. PA584852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPWPGQTSPV MAEYGTLLQD LTNNITLEDL GQLKSACKED IPSEKSEEIT TGSAWFSFLE SHNKLD Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 83%.

PEA15 is a death effector domain (DED)-containing protein predominantly expressed in the central nervous system, particularly in astrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEA15
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEA15
Gene Accession No. Q15121
Gene Alias 15 kDa phosphoprotein enriched in astrocytes; astrocytic phosphoprotein PEA-15; HMAT1; homolog of mouse MAT-1 oncogene; HUMMAT1H; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; MAT1; MAT1H; OTTHUMP00000029887; Pea15; PEA-15; Pea15a; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; phosphoprotein enriched in diabetes; Pkcs15; proliferation and apoptosis adaptor protein 15
Gene Symbols PEA15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEA15. Recombinant protein control fragment (Product #RP-108250).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8682
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.