Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEA15 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985469
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5469 Supplier Bioss Supplier No. BS3524R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PEA15 is a death effector domain (DED)-containing protein predominantly expressed in the central nervous system, particularly in astrocytes.

Specifications

Antigen PEA15
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PEA15
Gene Accession No. Q15121
Gene Alias 15 kDa phosphoprotein enriched in astrocytes; Astrocytic phosphoprotein PEA-15; BOS_2644; HMAT1; homolog of mouse MAT-1 oncogene; HUMMAT1H; LOC100355950; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; MAT1; MAT1H; OTTHUMP00000029887; Pea15; PEA-15; Pea15a; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; phosphoprotein enriched in astrocytes 15-like; Phosphoprotein enriched in diabetes; Pkcs15; proliferation and apoptosis adaptor protein 15
Gene Symbols PEA15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PEA15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8682
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.