Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEAK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191053
Description
PEAK1 Polyclonal specifically detects PEAK1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PEAK1 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| FLJ21140, FLJ34483, KIAA2002, NKF3 kinase family member, pseudopodium enriched atypical kinase, pseudopodium-enriched atypical kinase 1, SGK269, sugen kinase 269, tyrosine-protein kinase SgK269 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PEAK1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DGQSICGELSIQEHCENKPVIIGWNRNRAALSQKPLNNNNEDDEGISHVPKPYGNNDSAKKMSDNNNGLTEVLKEIAGLDTAPQIR | |
| 0.1 mL | |
| Protein Kinase | |
| 79834 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction