Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEAK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144707 Supplier Invitrogen™ Supplier No. PA5144707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMEKGRAENA LLQDSEKKRS HSSPSQIPKK ILSHMTHEVT EDFSPRDPRT VVGKQDGKGC TSVTTALSLP ELEREDGKED ISDPMDPNPC SATYSNLGQS RAAMIPPKQP Antigen sequence identity: Mouse 65%; Rat 60%.

Tyrosine kinase that may play a role in cell spreading and migration on fibronectin. May directly or indirectly affect phosphorylation levels of cytoskeleton-associated proteins MAPK1/ERK and PXN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEAK1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEAK1
Gene Accession No. Q9H792
Gene Alias 1110049L02Rik; 9530046P14; AA690169; BE132841; C230081A13Rik; Inactive tyrosine-protein kinase PEAK1; KIAA2002; mKIAA2002; NKF3 kinase family member; Peak1; pseudopodium enriched atypical kinase 1; pseudopodium-enriched atypical kinase 1; Sgk269; sugen kinase 269; Tyrosine-protein kinase SgK269
Gene Symbols PEAK1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1014-1123. Recombinant protein control fragment (Product #RP-109723).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.