Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEAK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555302 Supplier Invitrogen™ Supplier No. PA555302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGQSICGELS IQEHCENKPV IIGWNRNRAA LSQKPLNNNN EDDEGISHVP KPYGNNDSAK KMSDNNNGLT EVLKEIAGLD TAPQIR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.

Tyrosine kinase that may play a role in cell spreading and migration on fibronectin. May directly or indirectly affect phosphorylation levels of cytoskeleton-associated proteins MAPK1/ERK and PXN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEAK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEAK1
Gene Accession No. Q9H792
Gene Alias 1110049L02Rik; 9530046P14; AA690169; BE132841; C230081A13Rik; Inactive tyrosine-protein kinase PEAK1; KIAA2002; mKIAA2002; NKF3 kinase family member; Peak1; pseudopodium enriched atypical kinase 1; pseudopodium-enriched atypical kinase 1; Sgk269; sugen kinase 269; Tyrosine-protein kinase SgK269
Gene Symbols PEAK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEAK1. Recombinant protein control fragment (Product #RP-95222).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.