Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEAR1 Antibody (492621), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4527MFV450
Description
PEAR1 Monoclonal antibody specifically detects PEAR1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| PEAR1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 375033 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 492621 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| JEDI, platelet endothelial aggregation receptor 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human PEAR1, Leu21-Ser754, Accession # Q5VY43 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction