Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEAR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583262 Supplier Invitrogen™ Supplier No. PA583262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSDPNTCSFW ESFTTTTKES HSRPFSLLPS EPCERPWEGP HTCPQPTVVY RTVYRQVVKT DHRQRLQCCH GFYES Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 79%.

Platelet endothelial aggregation receptor 1 (PEAR1) is a platelet receptor that signals upon the formation of platelet-platelet contacts independent of platelet activation and secondary to platelet aggregation. Upon platelet aggregation stimulated by physiological agonists, PEAR1 becomes tyrosine- and serine-phosphorylated; the tyrosine phosphorylation can be inhibited by eptifibatide, an aIIbbeta3 antagonist that also inhibits platelet aggregation. Recent studies have indicated that genetic variations in PEAR1, may be associated with enhanced agonist-induced platelet aggregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEAR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEAR1
Gene Accession No. Q5VY43
Gene Alias 3110045G13Rik; hPEAR1; Jagged and Delta protein; Jedi; Jedi-1; MEGF12; mPEAR1; Multiple EGF-like domains protein 12; multiple EGF-like-domains 12; multiple epidermal growth factor-like domains protein 12; Pear1; Platelet endothelial aggregation receptor 1; protein Jedi; RGD1305653; RP11-107D16.3
Gene Symbols PEAR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEAR1. Recombinant protein control fragment (Product #RP-95727).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375033
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.