Learn More
PEAR1 Polyclonal Antibody

Description
PEAR1 Polyclonal Antibody for Western Blot, IHC
Specifications
Specifications
| Antigen | PEAR1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PEAR1 |
| Gene Accession No. | Q5VY43, Q8VIK5 |
| Gene Alias | 3110045G13Rik; hPEAR1; Jagged and Delta protein; Jedi; Jedi-1; MEGF12; mPEAR1; Multiple EGF-like domains protein 12; multiple EGF-like-domains 12; multiple epidermal growth factor-like domains protein 12; Pear1; Platelet endothelial aggregation receptor 1; protein Jedi; RGD1305653; RP11-107D16.3 |
| Gene Symbols | PEAR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.