Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEBP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589859 Supplier Invitrogen™ Supplier No. PA589859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEDENSPCAH EALLDEDTLF CQGLEVFYPE LGNIGCKVVP DCNNYRQKIT SWMEPIVKFP GAVDGATYIL VMVDPDAPSR AEPRQRFWRH WLVTDIKG; Positive Samples: Mouse lung; Cellular Location: Lysosome.

The phosphatidylethanolamine -binding proteins, including PEBP4, are an evolutionarily conserved family of proteins with pivotal biologic functions, such as lipid binding and inhibition of serine proteases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEBP4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pebp4
Gene Accession No. Q9D9G2
Gene Alias CORK1; CORK-1; cousin-of-RKIP 1 protein; epididymis secretory protein Li 300; GWTM1933; HEL-S-300; hPEBP4; MGC22776; PEBP4; PEBP-4; phosphatidylethanolamine binding protein 4; phosphatidylethanolamine-binding protein 4; PRO4408; Protein cousin-of-RKIP 1; UNQ1933/PRO4408
Gene Symbols Pebp4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 23-120 of human PEBP4 (NP_6593992).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73523
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.