Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1 Mouse anti-Human, PE, Clone: JC/70A, Abnova™

Catalog No. 89934755 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-755 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-755 Supplier Abnova Corporation Supplier No. MAB13014
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native PECAM1.

Specifications

Antigen PECAM1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone JC/70A
Conjugate PE
Description Mouse monoclonal antibody raised against native PECAM1.
Dilution Flow Cytometry (5 uL/million cells) Immunofluorescence (1:50-1:100 for 30 minutes at RT)
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Alias CD31/PECAM-1
Gene Symbols PECAM1
Host Species Mouse
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia.
Purification Method Protein A/G affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5175
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.