Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1 Rabbit anti-Human, Clone: RM247, Abnova™

Catalog No. 89035337 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-337 Supplier Abnova Corporation Supplier No. MAB15129
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against synthetic peptide of human PECAM1.

  • High specificity
  • High reactivity

Specifications

Antigen PECAM1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone RM247
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human PECAM1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:2500) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P16284
Gene Alias CD31/PECAM-1
Gene Symbols PECAM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to cytoplasmic domain region of human PECAM1.
Purification Method Protein A affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5175
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.