Learn More
3M™ Pecan Protein ELISA Kit, E96PEC, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products
Description
- Designed to detect processed and unprocessed target allergen proteins
- Test can accommodate environmental swab, CIP rinse water and food samples
- Universal extraction protocol streamlines testing for multiple analytes, saving reagents and time
Specifications
Specifications
| Detectable Analytes | Pecan protein |
| Product Type | ELISA kit |
| Quantity | 96-well plate |
| Test Time | 65 min. |