Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECI Monoclonal Antibody (OTI1C8), TrueMAB™, OriGene
SDP

Catalog No. 502540416
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0416 30 μL
50-167-4997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0416 Supplier OriGene Supplier No. TA504275S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PECI Monoclonal antibody specifically detects PECI in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PECI
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C8
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RFXANK
Gene Accession No. O14593
Gene Alias ACBD2; acyl-Coenzyme A binding domain containing 2; D3,D2-enoyl-CoA isomerase; DBI-related protein 1; delta(3),delta(2)-enoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; dJ1013A10.3; Dodecenoyl-CoA isomerase; Drs1; DRS-1; Eci2; enoyl-CoA delta isomerase 2; enoyl-CoA delta isomerase 2, mitochondrial; enoyl-Coenzyme A delta isomerase 2; Hca88; Hepatocellular carcinoma-associated antigen 88; PECI; Peroxisomal 3,2-trans-enoyl-CoA isomerase; peroxisomal 3,2-trans-enoyl-CoA isomerase; enoyl-CoA delta isomerase 2, mitochondrial; peroxisomal D3,D2-enoyl-CoA isomerase; peroxisomal delta3, delta2-enoyl-Coenzyme A isomerase; renal carcinoma antigen NY-REN-1; testicular secretory protein Li 33
Host Species Mouse
Immunogen Full length human recombinant protein of human RFXANK produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8625
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.