Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PECR Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297904AF750
Description
PECR Polyclonal specifically detects PECR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PECR | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
DCR-RP, EC 1.3.1.38, HPDHASE, HSA250303, peroxisomal trans-2-enoyl-CoA reductase, putative short chain alcohol dehydrogenase, pVI-ARL, PVIARL2,4-dienoyl-CoA reductase-related protein, SDR29C1, short chain dehydrogenase/reductase family 29C, member 1, TERPDCRRP | |
Produced in rabbits immunized with E. coli-derived Human PECR fragment. | |
0.1 mL | |
Apoptosis | |
55825 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction