Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PECR Monoclonal Antibody (OTI1C9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525262
Description
PECR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.
Specifications
PECR | |
Monoclonal | |
0.23 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BY49 | |
Pecr | |
Full length human recombinant protein of PECR produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI1C9 | |
Unconjugated | |
Pecr | |
2,4-dienoyl-CoA reductase-related protein; 2400003B18Rik; DCRRP; DCR-RP; HPDHase; HSA250303; PECR; perosisomal 2-enoyl-CoA reductase; peroxisomal 2,4-dienoyl-CoA reductase; Peroxisomal trans-2-enoyl-CoA reductase; PRO1004; putative short chain alcohol dehydrogenase; PVIARL; pVI-ARL; PX-2,4-DCR1; RLF98; SDR29C1; Short chain dehydrogenase/reductase family 29C member 1; short chain dehydrogenase/reductase family 29C, member 1; TERP | |
Mouse | |
Affinity chromatography | |
RUO | |
55825 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction