Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECR Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA501771

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-2890


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.
Specifications

Specifications

PECR
Monoclonal
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BY49, Q9WVK3
PECR
Full length human recombit protein of human PECR produced in HEK293T cell.
100 μL
Primary
Canine, Human, Rat
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2B2
Unconjugated
PECR
DCRRP, HPDHASE, HSA250303, PVIARL, SDR29C1, TERP
Mouse
Affinity Chromatography
RUO
113956, 478901, 55825
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.