Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PECR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554464 Supplier Invitrogen™ Supplier No. PA554464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRINCVAPGV IYSQTAVENY GSWGQSFFEG SFQKIPAKRI GVPEEVSSVV CFLLSPAASF ITGQSVDVDG GRSLYTHSYE VPDHDNWPKG AGDLSVVKKM KETFKEKAKL Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 63%.

PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PECR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pecr
Gene Accession No. Q9BY49
Gene Alias 2,4-dienoyl-CoA reductase-related protein; 2400003B18Rik; DCRRP; DCR-RP; HPDHase; HSA250303; PECR; perosisomal 2-enoyl-CoA reductase; peroxisomal 2,4-dienoyl-CoA reductase; Peroxisomal trans-2-enoyl-CoA reductase; PRO1004; putative short chain alcohol dehydrogenase; PVIARL; pVI-ARL; PX-2,4-DCR1; RLF98; SDR29C1; Short chain dehydrogenase/reductase family 29C member 1; short chain dehydrogenase/reductase family 29C, member 1; TERP
Gene Symbols Pecr
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PECR. Recombinant protein control fragment (Product #RP-93133).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55825
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.