Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pectinase from Aspergillus niger

Catalog No. ICN19897905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
10 kU
100 kU
25 kU
5 kU
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19897905 5 kU
ICN19897910 10 kU
ICN19897925 25 kU
ICN19897980 100 kU
ICN19897901 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19897905 Supplier MP Biomedicals™ Supplier No. 0219897905

Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.

  • Lyophilized
  • Activity: >1000 u/gm

TRUSTED_SUSTAINABILITY

Specifications

Concentration >1000 U/g
Color Beige
For Use With (Application) Used in plant protoplast preparation to digest cell wall prior to organelle isolation
Content And Storage 4°C
Source Aspergillus niger
Quantity 5 kU
Product Type Pectinase
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.