Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pectolyase Y-23 (A. Niger)

Catalog No. p-7163263
Click to view available options
Quantity:
1 g

Highly purified enzyme from Aspergillus niger used for isolation of biologically active protoplasts from a wide spectrum of higher plants and tissues.

  • Contains two types of pectinase such as endopolygalacturonase and endo-pectin lyase in high activity
  • Included is a maceration stimulating factor which stimulates tissue maceration by both pectinases
  • One unit is defined as an increase in A235 by 1.0 in the reaction mixture per minute
TRUSTED_SUSTAINABILITY

Specifications

Purity >1000 units per gram (pectin lyase)
Content And Storage Store at 2 to 8°C
Format Powder
For Use With (Application) Isolation of biologically active protoplasts
Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.