Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEDFR/PNPLA2/ATGL Rabbit anti-Human, Mouse, Rat, Clone: 10Y4M10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PEDFR/PNPLA2/ATGL |
---|---|
Clone | 10Y4M10 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
PEDFR/PNPLA2/ATGL Monoclonal antibody specifically detects PEDFR/PNPLA2/ATGL in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PEDFR/PNPLA2/ATGL | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Adipose triglyceride lipase, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF-R, Pigment epithelium-derived factor, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2.2, TTS-2.2, TTS2DKFZp667M109 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-394 of human PEDFR/PNPLA2/ATGL (Q96AD5). LPARLNEALLEACVEPTDLLTTLSNMLPVRLATAMMVPYTLPLESALSFTIRLLEWLPDVPEDIRWMKEQTGSICQYLVMRAKRKLGRHLPSRLP | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
10Y4M10 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
57104 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title