Learn More
Restek™ PEEK Finger-Tight Fitting
SDP
Catalog No. 06611830 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Pressure:
1500 psi
Catalog No. Pressure
06-611-830 1500 psi
1 options
Catalog No. 06-611-830 Supplier Restek™ Supplier No. 27849
For Tubing with 1/16 in. OD
  • Ideal for safely and securely connecting PTFE Tubing to Mobi-Cap Bottle Tops
  • Easily cut to desired length using Clean-Cut Tubing Cutter
Specifications
Type Fitting
Diameter (English) Outer 1/16 in.
For Use With (Application) Connecting PTFE Tubing to Mobi-Cap Bottle Tops
Thread Size 1/4-28
Pressure 1500 psi