Learn More
Excelta™ PEEK Tipped Tweezers
Catalog No. 18100826 Shop All Excelta Corporation Products
Change view
Click to view available options
Tip Style:
Curved
Catalog No. Tip Style
18-100-826 Curved
1 options
Catalog No. 18-100-826 Supplier Excelta™ Supplier No. 762ARTX
Add to Cart
Add to Cart
Highly resistant to chemicals and solvents
  • Inert to all common solvents
  • Widely used with semi-conductors
  • Working temperature of 249°C
Specifications
Product Type Replacement Tips
Material PEEK
Includes 2 tips and 4 screws
Quantity Each
Tip Style Curved