Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan Scientific and Medical PEEK Tubing, 1/32 in. O.D., 100 ft
SDP

Catalog No. 03350605 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (English) Inner:
0.0025 in.
0.004 in.
0.007 in.
0.010 in.
0.015 in.
0.020 in.
Color:
Black
Blue
Gray
Natural
Orange
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Inner Color
03-350-605 0.0025 in. Natural
03-350-606 0.004 in. Black
03-350-558 0.007 in. Yellow
03-350-559 0.010 in. Blue
03-350-560 0.015 in. Gray
03-350-561 0.020 in. Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-350-605 Supplier Trajan Scientific and Medical Supplier No. 1301002500100F

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs

Specifications

Color Natural
Diameter (English) Inner 0.0025 in.
Diameter (English) Outer 1/32 in.
Overall Length (English) 100 ft.
Material PEEK
Maximum Pressure 5000 psi (345 bar)
Max. Operating Temperature (Metric) 100°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.