Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan Scientific and Medical PEEK Tubing, 360μm I.D., 50 ft
SDP

Catalog No. 03350556 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (English) Inner:
125 μm (0.005 in.)
150 μm (0.006 in.)
50 μm (0.002 in.)
75 μm (0.003 in.)
Color:
Black
Green
Natural
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Inner Color
03-350-556 150 μm (0.006 in.) Yellow
03-350-552 50 μm (0.002 in.) Natural
03-350-553 75 μm (0.003 in.) Black
03-350-555 125 μm (0.005 in.) Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-350-556 Supplier Trajan Scientific and Medical Supplier No. 130400600250F

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs

Specifications

Color Yellow
pH Range 2 to 14
Diameter (English) Inner 150 μm (0.006 in.)
Diameter (Metric) Outer 360 μm
Overall Length (English) 50 ft.
Material PEEK
Maximum Pressure 2000 psi (138 bar)
Max. Operating Temperature (Metric) 100°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.