Learn More
Thermo Scientific™ Peezy Midstream Urine Collection Device
Description
Obtain a clean, midstream urine sample to support accurate urinalysis results with innovative and patient centric Peezy Midstream urine collection device.
Obtain a clean, midstream urine sample to support accurate urinalysis results with innovative and patient centric Peezy Midstream urine collection device.
The Peezy Midstream automatically captures a midstream sample by rejecting first void urine, capturing only the midstream, and when the container is full, rejecting the remainder of the urine stream. Minimal patient intervention and clinical explanation on how to provide a midstream sample are required.
A clean midstream urine sample is the gold standard1 for urinary tract infection diagnostics. The Peezy Midstream device’s improved collection method helps to reduce potential contaminants from a midstream urine sample to support the reduction of inaccurate or mixed culture results. Accurate diagnosis can facilitate appropriate treatment to promote improved antibiotic stewardship.
- Ease of use – Automatic midstream urine collection from an uninterrupted flow. No difficult or messy ‘start-stop-start’ required.
- Improved collection helps improve sample quality – Supports the reduction of culture contaminants from traditional ‘clean-catch’ urine pot collection.1
- Better sample could improve diagnostic workflow – May result in less retests, less false positives and a better sample-to-answer workflow.3
Peezy Midstream is available with an empty sample collection tube R649350) or with a pre-filled preservative (boric acid (0.18g)) (R649351) collection tube for the preservation of urine samples at room temperature.
Collection tubes are 95kPa leakproof with Secure screw cap, leak-tested in accordance with BS EN 14254. Suitable for centrifugation at 3,200 x g.
Specifications
Specifications
Product Type | Midstream Tube with Boric Acid |
Description | Peezy Misdtream (Boric acid tube) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.