Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584508 Supplier Invitrogen™ Supplier No. PA584508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPGSYYPGPP NSGGQYGSGL PPGGGYGGPA PGGPYGPPAG GGPYGHPNPG MFPSGTPGGP YGGAA Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 69%.

PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2. PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2 (PDCD6) (Kitaura et al., 2001 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEF1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEF1
Gene Accession No. Q9UBV8
Gene Alias 2600002E23Rik; ABP32; PEF protein with a long N-terminal hydrophobic domain; Pef1; PEF1A; Peflin; peflin; LOW QUALITY PROTEIN: peflin; penta-EF hand domain containing 1; penta-EF hand domain-containing protein 1; penta-EF-hand domain containing 1; UNQ1845/PRO3573; zgc:100787
Gene Symbols PEF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEF1. Recombinant protein control fragment (Product #RP-105196).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 553115
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.