Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114395
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114395 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114395 Supplier Invitrogen™ Supplier No. PA5114395
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human placenta tissue, human PC-3 whole cell, human U-87MG whole cell, human U-937 whole cell, human HepG2 whole cell, human A431 whole cell, human U2OS whole cell, rat lung tissues, rat testicular tissue, mouse lung tissue, mouse testicular tissue. IHC: mouse testis tissue, rat testis tissue, human colon cancer tissue, human lung cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2. PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2 (PDCD6) (Kitaura et al., 2001 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene PEF1
Gene Accession No. Q641Z8, Q8BFY6, Q9UBV8
Gene Alias 2600002E23Rik; ABP32; PEF protein with a long N-terminal hydrophobic domain; Pef1; PEF1A; Peflin; peflin; LOW QUALITY PROTEIN: peflin; penta-EF hand domain containing 1; penta-EF hand domain-containing protein 1; penta-EF-hand domain containing 1; UNQ1845/PRO3573; zgc:100787
Gene Symbols PEF1
Host Species Rabbit
Immunogen E.coli-derived human PEF1 recombinant protein (Position: R170-L284).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297900, 553115, 67898
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.