Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peflin Antibody (SR1512), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322558100UL
Description
Peflin Monoclonal antibody specifically detects Peflin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Peflin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1512 | |
Western Blot 1:500-1:2000 | |
ABP32, PEF protein with a long N-terminal hydrophobic domain, PEF1A, peflin, penta-EF hand domain containing 1, Penta-EF hand domain-containing protein 1, penta-EF-hand domain containing 1 | |
A synthesized peptide derived from human Peflin (Uniprot #: Q9UBV8) | |
100 μg | |
Signal Transduction | |
553115 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction