Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEG 35000, G-Biosciences
SCI_ED

Catalog No. 01001084
Click to view available options
Quantity:
5000 g, 1 mL
250 g
1000 g

Polyethylene glycol (PEG) is a polyether compound that is used as an additive in cosmetics, pharmaceuticals, and food. It consists of repeated ethylene glycol units[-(CH2CH2O)n].

PEG has several biomedical applications including biosensing, imaging, and tissue engineering. It also repels protein adsorption and lowers the liver uptake, which helps in coating nanoparticles.

Chemical Identifiers

Molecular Weight (g/mol) 35,000 g/mol

Specifications

Quantity 250 g
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder or Flakes
Product Line Gibco
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.