Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEG (methoxy group) Rabbit, Clone: RM105, Abnova™
Rabbit monoclonal antibody raised against polyethylene glycol (methoxy group).
Supplier: Abnova Corporation MAB12825
Specifications
| PEG (methoxy group) | |
| Monoclonal | |
| Unconjugated | |
| ELISA (0.01-0.3 ug/mL) Flow Cytometry Immunocytochemistry Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blot (0.05-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| Protein A affinity purification | |
| RUO | |
| This antibody reacts to the methoxy group of Polyethylene glycol (PEG). It is specific for methoxypoly (ethylene glycol). | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| RM105 | |
| Rabbit monoclonal antibody raised against polyethylene glycol (methoxy group). | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| Polyethylene glycol (methoxy group) conjugated with KLH. | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction