Learn More
Invitrogen™ PEG (methoxy group) Recombinant Rabbit Monoclonal Antibody (HL1020)

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Specifications
Specifications
| Antigen | PEG (methoxy group) |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | HL1020 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | Poly Ethylene Glycol; Polyethylene Glycol |
| Host Species | Rabbit |
| Immunogen | The immunogen used to generate this antibody corresponds to PEG (methoxy group). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.