Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEG10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $666.47
Specifications
| Antigen | PEG10 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PEG10 Polyclonal specifically detects PEG10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PEG10 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 23089 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:SIPNLEDQAHLATYTEFVPQIPGYQTYPTYAAYPTYPVGFAWYPVGRDGQGRSLYVPVMITWNPHWYRQPPVPQYPPPQPPPPPPPPPPPPSYSTL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| embryonal carcinoma differentiation regulated, Embryonal carcinoma differentiation-regulated protein, HB-1, KIAA1051EDR, Mammalian retrotransposon-derived protein 2, Mar2, Mart2, MEF3 like 1, MEF3L, MEF3L1, Myelin expression factor 3-like protein 1, paternally expressed 10, Paternally expressed gene 10 protein, retrotransposon gag domain containing 3, Retrotransposon gag domain-containing protein 3, Retrotransposon-derived gag-like polyprotein, retrotransposon-derived protein PEG10, RGAG3MEF3-like protein 1, Ty3/Gypsy-like protein | |
| PEG10 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title