Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEG10 Monoclonal Antibody (1B1C4)
GREENER_CHOICE

Catalog No. PIMA515475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515475 Supplier Invitrogen™ Supplier No. MA515475
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15475 targets PEG10 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15475 immunogen is purified recombinant fragment of PEG10 (aa1-120) expressed in E. Coli.

This is a paternally expressed imprinted gene that encodes transcripts containing two overlapping open reading frames start codon, have been reported for this gene. Increased expression of this gene is associated with hepatocellular carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEG10
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1B1C4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene PEG10
Gene Accession No. Q86TG7
Gene Alias 2-Mar; AA407948; EDR; embryonal carcinoma differentiation regulated; embryonal carcinoma differentiation regulated protein; Embryonal carcinoma differentiation-regulated protein; HB-1; KIAA1051; Mammalian retrotransposon-derived protein 2; MAR2; Mart2; MEF3 like 1; MEF3L; Mef3l1; MEF3-like protein 1; MyEF-3; MyEF-3 like; myelin expression factor 3; Myelin expression factor 3-like protein 1; paternally expressed 10; paternally expressed gene 10 protein; PEG10; Retrotransposon gag domain-containing protein 3; retrotransposon-derived gag-like polyprotein; Retrotransposon-derived protein PEG10; retrotransposon-derived protein PEG10-like; RGAG3; Ty3/Gypsy-like protein
Gene Symbols PEG10
Host Species Mouse
Immunogen Purified recombinant fragment of PEG10 (aa1-120) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.