Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEG10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562299 Supplier Invitrogen™ Supplier No. PA562299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QCIHIERRLA RAAAARKPRS PPRALVLPHI ASHHQVDPTE PVGGARMRLT QEEKERRRKL NLCLYCGTGG HYADNCPAKA SKSS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 28%.

This is a paternally expressed imprinted gene that encodes transcripts containing two overlapping open reading frames start codon, have been reported for this gene. Increased expression of this gene is associated with hepatocellular carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEG10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEG10
Gene Accession No. Q86TG7
Gene Alias 2-Mar; AA407948; EDR; embryonal carcinoma differentiation regulated; embryonal carcinoma differentiation regulated protein; Embryonal carcinoma differentiation-regulated protein; HB-1; KIAA1051; Mammalian retrotransposon-derived protein 2; MAR2; Mart2; MEF3 like 1; MEF3L; Mef3l1; MEF3-like protein 1; MyEF-3; MyEF-3 like; myelin expression factor 3; Myelin expression factor 3-like protein 1; paternally expressed 10; paternally expressed gene 10 protein; PEG10; Retrotransposon gag domain-containing protein 3; retrotransposon-derived gag-like polyprotein; Retrotransposon-derived protein PEG10; retrotransposon-derived protein PEG10-like; RGAG3; Ty3/Gypsy-like protein
Gene Symbols PEG10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEG10. Recombinant protein control fragment (Product #RP-100578).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.