Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEG10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501729430 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9430 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9430 Supplier Proteintech Group Inc Supplier No. 144121AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This is a paternally expressed imprinted gene that encodes transcripts containing two overlapping open reading frames start codon, have been reported for this gene. Increased expression of this gene is associated with hepatocellular carcinomas.

Specifications

Antigen PEG10
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PEG10
Gene Accession No. Q7TN75, Q86TG7
Gene Alias Edr, HB 1, KIAA1051, Mar2, Mart2, MEF3 like protein 1, MEF3L, MEF3L1, paternally expressed 10, PEG10, RGAG3, Ty3/Gypsy like protein
Gene Symbols Peg10
Host Species Rabbit
Immunogen PEG10 Fusion Protein Ag5754
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170676, 23089
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.