Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555282 Supplier Invitrogen™ Supplier No. PA555282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSNVREYQKA RAKKKYIEHR SNETSVIHSL PFGEQTFRPR GMLYECQECG ECFAHSSDLT EHQKIHDREK PSGSRNYEWS VIRSLAPTDP QTSYAQEQYA KEQARNKCKD FRQFFATSED LNTNQKIYDQ Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 69%.

In human, ZIM2 and PEG3 are treated as two distinct genes though they share multiple 5' exons and a common promoter and both genes are paternally expressed. Alternative splicing events connect their shared 5' exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. In contrast, in other mammals ZIM2 does not undergo imprinting and, in mouse, cow, and likely other mammals as well, the ZIM2 and PEG3 genes do not share exons. Human PEG3 protein belongs to the Kruppel C2H2-type zinc finger protein family. PEG3 may play a role in cell proliferation and p53-mediated apoptosis. PEG3 has also shown tumor suppressor activity and tumorigenesis in glioma and ovarian cells. Alternative splicing of this PEG3 gene results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEG3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEG3
Gene Accession No. Q9GZU2
Gene Alias AL022617; ASF-1; End4; Gcap4; granule cell antiserum positive 4; KIAA0287; Kruppel-type zinc finger protein; mKIAA0287; paternally expressed 3; paternally expressed gene 3; paternally-expressed gene 3 protein; PEG3; Pw1; Zfp102; zinc finger and SCAN domain-containing protein 24; ZKSCAN22; ZNF904; ZSCAN24
Gene Symbols PEG3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEG3. Recombinant protein control fragment (Product #RP-89445).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.