Learn More
Eisco™ Pegmatite Igneous Rock Specimen
SCI_ED
Catalog No. S27612 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27612 12/Pk.
S27590 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27612 Supplier Eisco™ Supplier No. ESNG0083PK12
Add to Cart
Add to Cart
Pegmatite is an igneous rock that contains two vastly different crystal sizes. This combination of both phaneritic and aphanitic crystals indicates this rock formed at two vastly different cooling rates.
  • Correlates well to Next Generation Science Standards Evidence
  • Pegmatite is a classic rock for teaching igneous petrology
  • Useful in volcanology lessons
Specifications
Product Type Igneous Rock
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov