Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pejvakin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116276
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116276 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116276 Supplier Invitrogen™ Supplier No. PA5116276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pejvakin Polyclonal Antibody for Western Blot

Essential in the activity of auditory pathway neurons. Defects in PJVK are the cause of non-syndromic sensorineural deafness autosomal recessive type 59 (DFNB59). DFNB59 is a form of sensorineural hearing impairment with absent or severely abnormal auditory brainstem response but normal otoacoustic emissions (auditory neuropathy or auditory dys-synchrony). Auditory neuropathies result from a lesion in the area including the inner hair cells, connections between the inner hair cells and the cochlear branch of the auditory nerve, the auditory nerve itself and auditory pathways of the brainstem. 'Pejvak' means 'echo' in Persian.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pejvakin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PJVK
Gene Accession No. Q0ZLH3
Gene Alias Autosomal recessive deafness type 59 protein; autosomal recessive deafness type 59 protein homolog; deafness, autosomal recessive 59; deafness, autosomal recessive 59 (human); Dfnb59; DFNB59 deafness; Gm1001; Pejvakin; Pjvk
Gene Symbols PJVK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 170-197 amino acids from the Central region of human PJVK.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494513
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.