Learn More
PELI1 Mouse anti-Human, Clone: OTI2H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807519

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Pellino proteins are a highly homologous family of E3 ubiquitin ligases that act as upstream mediators in Toll-like receptor (TLR) pathways that lead to activation of MAP kinases and transcription factors. Pellino 1 is required for interleukin-1-mediated signaling through its interaction with the IRAK4-IRAK-TRAF6 complex, ultimately resulting in the activation of NF-Kappa-B. Like other members of the Pellino family, Pellino 1 is an E3 ubiquitin ligase, able to catalyze the polyubiquitination of IRAK1. It is activated via phosphorylation by either IRAK1 and IRAK4 or the IKK-related kinases IKK-epsilon; and TBK1. In addition to phosphorylation, Pellino 1 activity is also modulated via ubiquitination and sumoylation.Specifications
PELI1 | |
Monoclonal | |
Unconjugated | |
PELI1 | |
pellino E3 ubiquitin protein ligase 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57162 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96FA3 | |
PELI1 | |
Human recombit protein fragment corresponding to amino acids 1-245 of human PELI1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.