Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pellino 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598795 Supplier Invitrogen™ Supplier No. PA598795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pellino 1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The Pellino proteins are a highly homologous family of E3 ubiquitin ligases that act as upstream mediators in Toll-like receptor (TLR) pathways that lead to activation of MAP kinases and transcription factors. Pellino 1 is required for interleukin-1-mediated signaling through its interaction with the IRAK4-IRAK-TRAF6 complex, ultimately resulting in the activation of NF-Kappa-B. Like other members of the Pellino family, Pellino 1 is an E3 ubiquitin ligase, able to catalyze the polyubiquitination of IRAK1. It is activated via phosphorylation by either IRAK1 and IRAK4 or the IKK-related kinases IKK-epsilon; and TBK1. In addition to phosphorylation, Pellino 1 activity is also modulated via ubiquitination and sumoylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pellino 1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PELI1
Gene Accession No. Q8C669, Q96FA3
Gene Alias 2810468L03Rik; A930031K15Rik; AA409794; AI586297; D11Ertd676e; E3 ubiquitin-protein ligase pellino homolog 1; Peli1; pellino 1; pellino E3 ubiquitin protein ligase 1; pellino homolog 1; pellino1; pellino-1; Pellino-related intracellular-signaling molecule; PRISM; protein pellino homolog 1; RING-type E3 ubiquitin transferase pellino homolog 1
Gene Symbols PELI1
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase pellino homolog 1 protein (1-250AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57162, 67245
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.