Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pellino 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564012 Supplier Invitrogen™ Supplier No. PA564012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSRLALSRRS HANGVKPDVM HHISTPLVSK ALSNRGQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Pellino 1 is the mammalian homologue of Pellino, a Drosophila Pelle-interacting protein. Pelle is a serine-threonine kinase through which Toll signaling can occur. Pellino 1 can activate c-Jun, Elk-1 and NF-kB, whereas Pellino 3 can only activate c-Jun and Elk-1(2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pellino 3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PELI3
Gene Accession No. Q8N2H9
Gene Alias 6030441F14Rik; A930011L17; BC028931; E3 ubiquitin-protein ligase pellino homolog 3; E3 ubiquitin-protein ligase pellino homolog 3 pseudogene; MGC35521; PELI3; pellino 3; pellino E3 ubiquitin protein ligase family member 3; pellino homolog 3; pellino-3; protein pellino homolog 3; RGD1305989; RING-type E3 ubiquitin transferase pellino homolog 3
Gene Symbols PELI3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Pellino 3. Recombinant protein control fragment (Product #RP-103442).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246330
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.