Learn More
Invitrogen™ PELO Polyclonal Antibody

Description
Immunogen sequence: KANEAMAIDT LLISDELFRH QDVATRSRYV RLVDSVKENA GTVRIFSSLH VSGEQLSQLT GVAAILRFPV PELSDQEGDS SSEED Antigen sequence identity: Mouse 95%; Rat 95%.
Specifications
Specifications
| Antigen | PELO |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PELO |
| Gene Accession No. | Q9BRX2 |
| Gene Alias | AA409897; AI852218; CGI-17; PELO; pelota homolog (Drosophila); PRO1770; Protein pelota homolog |
| Gene Symbols | PELO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.