Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PELO Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502208450
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8450 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8450 Supplier Fabgennix Inc Supplier No. PELO101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PELO Polyclonal antibody specifically detects PELO in Human, Mouse, Non-human Primate, Rat samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen PELO
Applications Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PELO
Gene Accession No. Q5XIP1, Q80X73, Q9BRX2
Gene Alias AA409897; AI852218; CGI-17; PELO; pelota homolog (Drosophila); PRO1770; Protein pelota homolog
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 330-380 on human PELOTA protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104001088, 105083, 294754, 53918
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.