Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PELO Recombinant Rabbit Monoclonal Antibody (HL2345)
GREENER_CHOICE

Catalog No. FEMA550497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550497 Supplier Invitrogen™ Supplier No. MA550497
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

PELO Recombinant Monoclonal Antibody for Western Blot, IHC, IHC (P)

PELO encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PELO
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2345
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PELO
Gene Accession No. Q5XIP1, Q80X73, Q9BRX2
Gene Alias AA409897; AI852218; CGI-17; PELO; pelota homolog (Drosophila); PRO1770; Protein pelota homolog
Gene Symbols PELO
Host Species Rabbit
Immunogen Recombinant fragment of human PELO.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105083, 294754, 53918
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.