Learn More
3M™ PELTOR™ FLX2 Cable for CH-3 Listen Only Headset
PELTOR™ FLX2 platform allows workers to pair protective communication CH-3 listen-only headsets with existing handheld devices.
Supplier: 3M™ 7100191444
Description
- For use with CH-3 Listen-Only Headsets
- Simple and reliable connection to a wide variety of handheld devices.
- Coiled polyurethane cable
- Aramid fiber reinforced
- Robust and highly flexible
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Black | |
Enables connection to listen-only CH-3 Headsets with Hytera, Icom, Kenwood or Motorola radios | |
1 Ea. |
1/4 in. stereo plug | |
FLX2-204 | |
FLX2 Cable |